missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Morpholinoisonicotinaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 2-morpholinoisonicotinaldehyde, 2-morpholin-4-yl pyridine-4-carbaldehyde, 2-morpholin-4-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-4-morpholinyl, 2-morpholin-4-yl isonicotinaldehyd, 2-morpholin-4-yl-pyridine-4-carbaldehyde, 4-pyridinecarboxaldehyde, 2-4-morpholinyl |
IUPAC Name | 2-(morpholin-4-yl)pyridine-4-carbaldehyde |
Formula Weight | 192.22 |
Percent Purity | 97% |
Chemical Name or Material | 2-Morpholinoisonicotinaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'