missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinobenzylamine, 97%, Thermo Scientific™

Catalog Number p-2698680
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
204078-48-8
C11H16N2O
192.26
MFCD03086183
RNYURNUANACIKS-UHFFFAOYSA-N
2-morpholinobenzylamine, 2-morpholinophenyl methanamine, 2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-yl-benzylamine, 2-morpholin-4-yl benzylamine, 2-4-morpholinyl phenyl methanamine, 1-2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-ylphenyl methylamine, 2-morpholino benzylamine, 2-morpholin-4-ylbenzylamine
2776563
(2-morpholin-4-ylphenyl)methanamine
NCC1=CC=CC=C1N1CCOCC1
This item is not returnable. View return policy
204078-48-8
C11H16N2O
192.26
MFCD03086183
RNYURNUANACIKS-UHFFFAOYSA-N
2-morpholinobenzylamine, 2-morpholinophenyl methanamine, 2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-yl-benzylamine, 2-morpholin-4-yl benzylamine, 2-4-morpholinyl phenyl methanamine, 1-2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-ylphenyl methylamine, 2-morpholino benzylamine, 2-morpholin-4-ylbenzylamine
2776563
(2-morpholin-4-ylphenyl)methanamine
NCC1=CC=CC=C1N1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 2-Morpholinobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'