missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Methylthio)thiophene, 97%

Brand:  Alfa Aesar™ A11763.14

Code : NEW

Additional Details : CAS Number : 5780-36-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11479323

  • £435.00 / 25g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Description

Description

Applications
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2- (Methylthio)thiophene
C5H6S2
MFCD00052382
106272
5780-36-9
130.23
UN3334
2-(Methylmercapto)thiophene; Methyl 2-thienyl sulfide

Specifications

2- (Methylthio)thiophene
82° to 84°C (15mmHg)
5780-36-9
C5H6S2
MFCD00052382
106272
Stench
25g
97%
130.23
UN3334
2-(Methylmercapto)thiophene; Methyl 2-thienyl sulfide
Videos
SDS