missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals

Catalog Number p-4259038
38.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


34.50 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.


Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
96-47-9
C5H10O
86.13
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
7301
2-methyloxolane
CC1CCCO1
96-47-9
C5H10O
86.13
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
7301
2-methyloxolane
CC1CCCO1

2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, ACROS Organics, Quantity: 10L, Packaging: Metal Drum, Boiling Point: 78.0 deg.C to 80.0 deg.C, CAS: 96-47-9, Colorless, Grade: For Grignard reaction, Melting Point: -136.0 deg.C, Molecular Formula: C5H10O, Molecular Weight: 86.13

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -136.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 78.0°C to 80.0°C
Quantity 250 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4046 to 1.4066
Specific Gravity 0.86
Viscosity 4 mPa.s (25°C)
Merck Index 15,6198
Solubility Information Solubility in water: 150g/l (25°C)
Formula Weight 86.13
Percent Purity ≥99.0%
Grade For Grignard reaction
Chemical Name or Material 2-Methyltetrahydrofuran, Stabilized; For Grignard reaction
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyltetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'