missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran-3-thiol, 97%, mixed isomers, ACROS Organics™

Brand:  Acros Organics 332700010

Code : 29

Additional Details : CAS Number : 57124-87-5 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10747592

  • £17.00 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

2-Methyltetrahydrofuran-3-thiol
57124-87-5
100.0
97%
118.2
Mixture of isomers
96.0
97%
C5H10OS
MFCD01632596

Specifications

2-Methyltetrahydrofuran-3-thiol
1.04
Clear colorless to pale yellow
Mixture of isomers
96.0
1.0400g/mL
Glass bottle
Liquid
97%
118.2
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
40°C
160.0° to 180.0°C
1g
57124-87-5
100.0
Authentic
1.4730 to 1.4910
97%
C5H10OS
MFCD01632596
Videos
SDS