missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltetrahydrofuran-3-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Formula Weight | 100.12 |
Percent Purity | 97% |
Density | 1.045g/mL |
Quantity | 1 g |
Packaging | Amber Glass Bottle |
Chemical Name or Material | 2-Methyltetrahydrofuran-3-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'