missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran-3-one, 97%, Maybridge

Brand:  Maybridge SB01113EA

Code : 31

Additional Details : CAS Number : 3188-00-9 Weight : 0.06000kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10482634

  • £33.00 / 10g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyltetrahydrofuran-3-one
97.0
97%
100.12
Dihydro-2-methyl-3(2H)-furanone
3188-00-9
97%
C5H8O2
MFCD00010423

Specifications

2-Methyltetrahydrofuran-3-one
3188-00-9
1.045g/mL
97%
C5H8O2
MFCD00010423
10g
97.0
Amber Glass Bottle
97%
100.12
Dihydro-2-methyl-3(2H)-furanone
Videos
SDS