missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran-3-one, 97%, Thermo Scientific™

Catalog Number p-2698305
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
3188-00-9
C5H8O2
100.117
MFCD00010423
FCWYQRVIQDNGBI-UHFFFAOYSA-N
2-methyltetrahydrofuran-3-one, 2-methyl tetrahydro-3-furanone, 2-methyltetrahydro-3-furanone, 3 2h-furanone, dihydro-2-methyl, dihydro-2-methyl-3 2h-furanone, coffee furanone, dihydro-2-methyl-3-furanone, tetrahydro-2-methylfuran-3-one, 2-methyl-3-oxotetrahydrofuran, 2-methyl-3-ketotetrahydrofuran
18522
2-methyloxolan-3-one
CC1C(=O)CCO1
This item is not returnable. View return policy
3188-00-9
C5H8O2
100.117
MFCD00010423
FCWYQRVIQDNGBI-UHFFFAOYSA-N
2-methyltetrahydrofuran-3-one, 2-methyl tetrahydro-3-furanone, 2-methyltetrahydro-3-furanone, 3 2h-furanone, dihydro-2-methyl, dihydro-2-methyl-3 2h-furanone, coffee furanone, dihydro-2-methyl-3-furanone, tetrahydro-2-methylfuran-3-one, 2-methyl-3-oxotetrahydrofuran, 2-methyl-3-ketotetrahydrofuran
18522
2-methyloxolan-3-one
CC1C(=O)CCO1
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 100.12
Percent Purity 97%
Density 1.045g/mL
Quantity 1 g
Packaging Amber Glass Bottle
Chemical Name or Material 2-Methyltetrahydrofuran-3-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'