missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)acetonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-4229311
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2274-42-2
C3H5NO2S
119.14
FOTRKCAZUSJCQD-UHFFFAOYSA-N
methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
75283
CHEBI:32444
2-methylsulfonylacetonitrile
CS(=O)(=O)CC#N
This item is not returnable. View return policy
2274-42-2
C3H5NO2S
119.14
FOTRKCAZUSJCQD-UHFFFAOYSA-N
methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
75283
CHEBI:32444
2-methylsulfonylacetonitrile
CS(=O)(=O)CC#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 86.0°C
Color White
Elemental Analysis 29.03 to 31.45% C, 11.29 to 12.23% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 119.14
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-(Methylsulfonyl)acetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Methylsulfonyl)acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'