missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)acetonitrile, 97%, ACROS Organics™

Brand:  Acros Organics 438420250

Code : 29

Additional Details : CAS Number : 2274-42-2 Weight : 0.07500kg Transport : UN number : 3439 Chem class : 1 Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11915761

  • £139.90 / 25g


Estimated Shipment
21-05-2018
Add to basket


Specifications

Chemical Identifiers

2-(Methylsulfonyl)acetonitrile
96.0
97%
119.14
2274-42-2
100.0
C3H5NO2S

Specifications

2-(Methylsulfonyl)acetonitrile
83.0° to 86.0°C
2274-42-2
100.0
Authentic
Powder or Crystalline Powder
97%
119.14
White to off-white
25g
96.0
29.03 to 31.45% C, 11.29 to 12.23% N
Glass bottle
97%
C3H5NO2S
Videos
SDS