missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrrolidine, 98%, Thermo Scientific Chemicals

Catalog Number p-2690117
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
765-38-8
C5H11N
85.15
MFCD00014491
RGHPCLZJAFCTIK-UHFFFAOYSA-N
13003
2-methylpyrrolidine
CC1CCCN1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
765-38-8
C5H11N
85.15
MFCD00014491
RGHPCLZJAFCTIK-UHFFFAOYSA-N
13003
2-methylpyrrolidine
CC1CCCN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 98°C to 100°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.437
Quantity 2 g
Specific Gravity 0.88
Synonym pyrrolidine, 2-methyl, 2-methyl-pyrrolidine, 2-methylpyrolidine, 2-methyl-pyrolidine, 2-methyl pyrrolidine, pubchem9410, pyrrolidine,2-methyl, acmc-209mch, acmc-1aspi, rac-2-methyl-pyrrolidine
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in common organic solvents
Formula Weight 85.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylpyrrolidine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methylpyrrolidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'