missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrazine, 99+%, Thermo Scientific Chemicals

Catalog Number p-167445
15.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
109-08-0
C5H6N2
94.12
MFCD00006142
CAWHJQAVHZEVTJ-UHFFFAOYSA-N
methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
7976
2-methylpyrazine
CC1=NC=CN=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
109-08-0
C5H6N2
94.12
MFCD00006142
CAWHJQAVHZEVTJ-UHFFFAOYSA-N
methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
7976
2-methylpyrazine
CC1=NC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -29.0°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 134.0°C to 137.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5042
Quantity 25 g
Beilstein 23, 94
Specific Gravity 1.03
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, ether, acetone, oils and, halogenated solvents
Formula Weight 94.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methylpyrazine, 99+ %
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'