missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnicotinic acid, 98+%, ACROS Organics™

£54.30 - £670.00

Chemical Identifiers

CAS 3222-56-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00013449
InChI Key HNTZKNJGAFJMHQ-UHFFFAOYSA-N
Synonym 2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
PubChem CID 643373
IUPAC Name 2-methylpyridine-3-carboxylic acid
SMILES CC1=C(C=CC=N1)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10061594
SDS View Documents
Acros Organics
397320010
1g
£54.30
1g
View alternative products
Save Up to 
»
Estimated Shipment: 14-10-2021
Please to purchase this item. Need a web account? Register with us today!
10071594
SDS View Documents
Acros Organics
397320050
5g
£206.00
5g
View alternative products
Save Up to 
»
Estimated Shipment: 07-10-2021
Please to purchase this item. Need a web account? Register with us today!
11926341
SDS View Documents
Acros Organics
397320250
25g
£670.00
25g
View alternative products
Save Up to 
»
Estimated Shipment: 06-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

3222-56-8
137.14
HNTZKNJGAFJMHQ-UHFFFAOYSA-N
643373
CC1=C(C=CC=N1)C(=O)O
C7H7NO2
MFCD00013449
2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
2-methylpyridine-3-carboxylic acid

Specifications

2-Methylnicotinic acid
Glass bottle
225.0°C to 228.0°C
3222-56-8
100.0
C7H7NO2
2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
CC1=C(C=CC=N1)C(=O)O
137.14
137.14
98+%
Authentic
White to Brown
1g
98.0
98% min. (HPLC)
MFCD00013449
HNTZKNJGAFJMHQ-UHFFFAOYSA-N
2-methylpyridine-3-carboxylic acid
643373
Powder or Crystalline
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methylnicotinic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Product Certifications