missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnaphthalene, 96%, Thermo Scientific Chemicals

Catalog Number p-165011
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
91-57-6
C11H10
142.2
MFCD00004118
QIMMUPPBPVKWKM-UHFFFAOYSA-N
naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10
7055
CHEBI:50720
2-methylnaphthalene
CC1=CC2=CC=CC=C2C=C1
This item is not returnable. View return policy
91-57-6
C11H10
142.2
MFCD00004118
QIMMUPPBPVKWKM-UHFFFAOYSA-N
naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10
7055
CHEBI:50720
2-methylnaphthalene
CC1=CC2=CC=CC=C2C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96
Melting Point 32°C to 36°C
Color White
Density 1.0100g/mL
Boiling Point 241°C to 242°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH3
Quantity 5 g
Beilstein 05, 567
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 142.2
Percent Purity 96%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Methylnaphthalene, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'