missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole-d6, TRC

Product Code. 30180239
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180239 5 mg 5mg
30180204 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30180239 Supplier Toronto Research Chemicals Supplier No. TRCM3129865MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1173022-19-9
Molecular Formula C4 D6 N2
Molecular Weight (g/mol) 88.1408
Synonym 1H-Imidazole-1,4,5-d3, 2-(methyl-d3)- (ACI), 2-(Methyl-d3)-1H-imidazole-1,4,5-d3 (ACI), 2-Methylimidazole-d6, 2-Methyl-1H-imidazole-D6
IUPAC Name 1,4,5-trideuterio-2-(trideuteriomethyl)imidazole
SMILES [2H]c1nc(n([2H])c1[2H])C([2H])([2H])[2H]

Spezifikation

Quantity 5 mg
InChI Formula InChI=1S/C4H6N2/c1-4-5-2-3-6-4/h2-3H,1H3,(H,5,6)/i1D3,2D,3D/hD
Formula Weight 88.0908
Chemical Name or Material 2-Methylimidazole-d6
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.