missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7081354
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4536-23-6
C7H13O2
129.18
MFCD00002674
CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl
20653
CCCC[C@@H](C)C([O-])=O
This item is not returnable. View return policy
4536-23-6
C7H13O2
129.18
MFCD00002674
CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl
20653
CCCC[C@@H](C)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.916
Boiling Point 209°C to 210°C
Flash Point 105°C (221°F)
Odor Rancid
Refractive Index 1.4205
Quantity 5 g
UN Number UN3265
Beilstein 1721227
IUPAC Name 2-methylhexanoic acid
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 2-Methylhexanoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'