missing translation for 'onlineSavingsMsg'
Learn More

2-Methylene-1,3-propanediol, 97%, Thermo Scientific Chemicals

Catalog Number p-3758394
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3513-81-3
C4H8O2
88.11
MFCD00075162
JFFYKITVXPZLQS-UHFFFAOYSA-N
2-methylene-1,3-propanediol, 2-methylenepropane-1,3-diol, 1,3-propanediol, 2-methylene, 1,3-dihydroxy-2-methylenepropane, 1,3-dihydroxy-2-methylidenepropane, dimethylol ethylene, 2-methylene1,3-propanediol, 2-methylenepropan-1,3-diol, ksc573o4t, 1,3-propanediol,2-methylene
77048
2-methylidenepropane-1,3-diol
C=C(CO)CO
This item is not returnable. View return policy
3513-81-3
C4H8O2
88.11
MFCD00075162
JFFYKITVXPZLQS-UHFFFAOYSA-N
2-methylene-1,3-propanediol, 2-methylenepropane-1,3-diol, 1,3-propanediol, 2-methylene, 1,3-dihydroxy-2-methylenepropane, 1,3-dihydroxy-2-methylidenepropane, dimethylol ethylene, 2-methylene1,3-propanediol, 2-methylenepropan-1,3-diol, ksc573o4t, 1,3-propanediol,2-methylene
77048
2-methylidenepropane-1,3-diol
C=C(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 93°C to 95°C (2.0 mmHg)
Flash Point <110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2C(=CH2)CH2OH
Refractive Index 1.4730 to 1.475
Quantity 1 g
Specific Gravity 1.08
Formula Weight 88.11
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 2-Methylene-1, 3-propanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'