2-Methylbutane, +99%, ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 397220010

1LT 2-Methylbutane, 99+%, Extra Dry, AcroSeal

UNSPSC: 12352100

Code: 50

Additional Details:
Additional Details: CAS Number: 78-78-4 Weight: 0.67000kg Transport: UN number : 1265 Chem class : 1 Pack group : I


Packaging AcroSeal™ Glass Bottle
Quantity 1L

Product Code. 10510384

Quantity Price
1 £ 65.0 / 1 L
Estimated Shipment
20-02-2017
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Methylbutane
Name Note Extra Dry
CAS 78-78-4
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Extra Dry
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C5H12
Linear Formula C2H5CH(CH3)2
Formula Weight 72.15
MDL Number MFCD00009338
Synonym Isopentane

Specification

Assay ≥99.0%
Physical Form Liquid
Formula Weight 72.15
Quantity 1L
Color Clear colorless
Packaging AcroSeal™ Glass Bottle
CAS Min % 99.0
Residue after Evaporation 0.001% max.
Linear Formula C2H5CH(CH3)2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Purity Grade Extra Dry
CAS 78-78-4
Assay Percent Range 99+%
MDL Number MFCD00009338
Water 0.002% max.
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Density 0.62g/mL
Name Note Extra Dry
Molecular Formula C5H12
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Chemical Name or Material 2-Methylbutane
Synonym Isopentane
Documentation Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.