missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzophenone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022148
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
131-58-8
C14H12O
196.249
MFCD00008518
CKGKXGQVRVAKEA-UHFFFAOYSA-N
2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone
67230
(2-methylphenyl)-phenylmethanone
CC1=CC=CC=C1C(=O)C2=CC=CC=C2
This item is not returnable. View return policy
131-58-8
C14H12O
196.249
MFCD00008518
CKGKXGQVRVAKEA-UHFFFAOYSA-N
2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone
67230
(2-methylphenyl)-phenylmethanone
CC1=CC=CC=C1C(=O)C2=CC=CC=C2

2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressure.
TRUSTED_SUSTAINABILITY
Melting Point -18°C
Density 1.085
Boiling Point 309°C to 311°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 10 g
Beilstein 2045469
Merck Index 14,7317
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'