missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanisole, 99%, ACROS Organics™

Brand:  Acros Organics 126280050

Code : 29

Additional Details : CAS Number : 578-58-5 Weight : 0.05500kg Transport : UN number : 1993 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10523291

  • £10.20 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

2-Methylanisole
578-58-5
100.0
99%
CH3C6H4OCH3
MFCD00008373
99%
98.5
≥98.5%
C8H10O
122.17
1-Methoxy-2-methylbenzene

Specifications

2-Methylanisole
Clear colorless to light yellow
99%
98.5
Authentic
Glass bottle
≥98.5%
C8H10O
122.17
06, 352
0.9800g/mL
5g
578-58-5
100.0
1.5160 to 1.5180 (20°C, 589nm)
Liquid
99%
CH3C6H4OCH3
MFCD00008373
1-Methoxy-2-methylbenzene
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methylanisole

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS