missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals (+)-2-Methyl-L-serine, 99%
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 16820-18-1 |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 286.0°C to 288.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C4H9NO3 |
Linear Formula | HOCH2C(CH3)NH2COOH |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'