missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl hydrogen pyridine-2,5-dicarboxylate, 97%, Maybridge

Brand:  Maybridge MO08350DE

Code : 31

Additional Details : CAS Number : 17874-76-9 Weight : 0.00500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 11356905

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Methyl hydrogen pyridine-2, 5-dicarboxylate
97.0
97%
181.15
17874-76-9
97%
C8H7NO4
MFCD09702182

Specifications

2-Methyl hydrogen pyridine-2, 5-dicarboxylate
17874-76-9
Amber Glass Bottle
97%
181.15
5g
97.0
97%
C8H7NO4
MFCD09702182
Videos
SDS