missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl hydrogen pyridine-2,5-dicarboxylate, 97%, Maybridge

Brand:  Maybridge MO08350DA

Code : 31

Additional Details : CAS Number : 17874-76-9 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10163364

  • £102.00 / 1g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl hydrogen pyridine-2, 5-dicarboxylate
97.0
97%
181.15
17874-76-9
97%
C8H7NO4
MFCD09702182

Specifications

2-Methyl hydrogen pyridine-2, 5-dicarboxylate
17874-76-9
Amber Glass Bottle
97%
181.15
1g
97.0
97%
C8H7NO4
MFCD09702182
Videos
SDS