missing translation for 'onlineSavingsMsg'
Learn More

(2-methyl-benzoimidazol-1-yl)-acetic acid, 97%, Maybridge™

Brand:  Maybridge MO08104CB

Code : 31

Additional Details : CAS Number : 40332-17-0 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 10001014

  • £24.11 / 250mg

Estimated Shipment
14-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

40332-17-0
190.202
SMGBDFGKVGSCGB-UHFFFAOYSA-N
719681
CC1=NC2=CC=CC=C2N1CC(=O)O
C10H10N2O2
MFCD00621731
2-methyl-1h-benzimidazol-1-yl acetic acid, 2-2-methyl-1h-benzo d imidazol-1-yl acetic acid, 2-methyl-benzoimidazol-1-yl-acetic acid, 2-methyl-1h-benzimidazole-1-acetic acid, 1h-benzimidazole-1-acetic acid, 2-methyl, 2-methylbenzoimidazol-1-yl acetic acid, 2-methyl-benzoimidazol-1-yl acetic acid, 2-2-methylbenzimidazol-1-yl acetic acid, 2-methyl-1,3-benzodiazol-1-yl acetic acid, 2-2-methylbenzimidazolyl acetic acid
2-(2-methylbenzimidazol-1-yl)acetic acid

Specifications

(2-Methyl-benzoimidazol-1-yl)-acetic acid
40332-17-0
97%
MFCD00621731
SMGBDFGKVGSCGB-UHFFFAOYSA-N
2-(2-methylbenzimidazol-1-yl)acetic acid
719681
97%
250mg
97.0
C10H10N2O2
2-methyl-1h-benzimidazol-1-yl acetic acid, 2-2-methyl-1h-benzo d imidazol-1-yl acetic acid, 2-methyl-benzoimidazol-1-yl-acetic acid, 2-methyl-1h-benzimidazole-1-acetic acid, 1h-benzimidazole-1-acetic acid, 2-methyl, 2-methylbenzoimidazol-1-yl acetic acid, 2-methyl-benzoimidazol-1-yl acetic acid, 2-2-methylbenzimidazol-1-yl acetic acid, 2-methyl-1,3-benzodiazol-1-yl acetic acid, 2-2-methylbenzimidazolyl acetic acid
CC1=NC2=CC=CC=C2N1CC(=O)O
190.202
190.2
Videos
SDS Documents