missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroaniline 99%, ACROS Organics™

Catalog Number p-164997
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
570-24-1
C7H8N2O2
152.153
MFCD00007744
FCMRHMPITHLLLA-UHFFFAOYSA-N
6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc
11298
2-methyl-6-nitroaniline
CC1=C(C(=CC=C1)[N+](=O)[O-])N
This item is not returnable. View return policy
570-24-1
C7H8N2O2
152.153
MFCD00007744
FCMRHMPITHLLLA-UHFFFAOYSA-N
6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc
11298
2-methyl-6-nitroaniline
CC1=C(C(=CC=C1)[N+](=O)[O-])N
CAS Min % 98.5
CAS Max % 100.0
Melting Point 93.0°C to 96.0°C
Color Brown
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 25 g
Beilstein 12, 843
Formula Weight 152.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-6-nitroaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-6-nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'