missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroaniline 99%, ACROS Organics™

Brand:  Acros Organics 127230050

Code : 29

Additional Details : CAS Number : 570-24-1 Weight : 0.05500kg Transport : UN number : 2660 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10711155

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Methyl-6-nitroaniline
570-24-1
100.0
99%
CH3C6H3(NO2)NH2
MFCD00007744
99%
98.5
99%
C7H8N2O2
152.15
2-Amino-3-nitrotoluene, 6-Nitro-o-toluidine

Specifications

2-Methyl-6-nitroaniline
570-24-1
100.0
Glass bottle
99%
C7H8N2O2
152.15
12, 843
2-Amino-3-nitrotoluene, 6-Nitro-o-toluidine
Solubility in water: negligible. Other solubilities: slouble in ethanol and ether
110°C
99%
98.5
Authentic
Crystalline Powder
99%
CH3C6H3(NO2)NH2
MFCD00007744
Brownish
93.0° to 96.0°C
5g
Videos
SDS