missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)benzonitrile, 97%, Thermo Scientific™

Catalog Number 15407077
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1261679-13-3
C9H6F3NO
201.15
MFCD18399750
VZVKYRYUNZHYII-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzonitrile
86277656
2-methyl-5-(trifluoromethoxy)benzonitrile
CC1=C(C=C(OC(F)(F)F)C=C1)C#N
This item is not returnable. View return policy
1261679-13-3
C9H6F3NO
201.15
MFCD18399750
VZVKYRYUNZHYII-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzonitrile
86277656
2-methyl-5-(trifluoromethoxy)benzonitrile
CC1=C(C=C(OC(F)(F)F)C=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3276
Formula Weight 201.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-5-(trifluoromethoxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'