missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-propionyl-furan, 95%, ACROS Organics™

Brand:  Acros Organics 233000100

Code : 29

Additional Details : CAS Number : 10599-69-6 Weight : 0.06000kg

Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10328610

  • £37.90 / 10g


Estimated Shipment
08-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-5-propionyl-furan
10599-69-6
100.0
95%
138.17
95%
94.0
95%
C8H10O2
MFCD00053142

Specifications

2-Methyl-5-propionyl-furan
95%
94.0
Authentic
1.5037 to 1.5057
95%
C8H10O2
MFCD00053142
10g
10599-69-6
100.0
Glass bottle
Liquid
95%
138.17
Clear yellow
Videos
SDS