missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furyl isocyanate, Thermo Scientific™

Catalog Number p-4510629
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
568577-82-2
C12H9NO2
199.209
LVNIEPBRJLDJHU-UHFFFAOYSA-N
2-methyl-5-phenyl-3-furyl isocyanate, 2-methyl-5-phenyl-3-furylisocyanate, 2-methyl-5-phenylfuran-3-isocyanate, 3-isocyanato-2-methyl-5-phenyl-furan
2795315
3-isocyanato-2-methyl-5-phenylfuran
CC1=C(C=C(O1)C2=CC=CC=C2)N=C=O
This item is not returnable. View return policy
568577-82-2
C12H9NO2
199.209
LVNIEPBRJLDJHU-UHFFFAOYSA-N
2-methyl-5-phenyl-3-furyl isocyanate, 2-methyl-5-phenyl-3-furylisocyanate, 2-methyl-5-phenylfuran-3-isocyanate, 3-isocyanato-2-methyl-5-phenyl-furan
2795315
3-isocyanato-2-methyl-5-phenylfuran
CC1=C(C=C(O1)C2=CC=CC=C2)N=C=O
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'