missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroimidazole, 99%, Thermo Scientific Chemicals

Catalog Number p-164985
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
88054-22-2
C4H5N3O2
127.10
MFCD00005191
FFYTTYVSDVWNMY-UHFFFAOYSA-N
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
12760
2-methyl-5-nitro-1H-imidazole
CC1=NC=C(N1)[N+]([O-])=O
This item is not returnable. View return policy
88054-22-2
C4H5N3O2
127.10
MFCD00005191
FFYTTYVSDVWNMY-UHFFFAOYSA-N
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
12760
2-methyl-5-nitro-1H-imidazole
CC1=NC=C(N1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 252°C to 254°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 50 g
Beilstein 23, I, 23
Formula Weight 127.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Methyl-5-nitroimidazole, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'