missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-5-nitrobenzotrifluoride, 98%

Brand:  Alfa Aesar™ H64924.03

Code : NEW

Additional Details : CAS Number : 89976-12-5 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15432349

  • £14.00 / 1g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-Methyl-5-nitrobenzotrifluoride
205.14
UN2811
1-Methyl-4-nitro-2-(trifluoromethyl)benzene
89976-12-5
MFCD01631684
2506013

Specifications

2-Methyl-5-nitrobenzotrifluoride
Solid
205.14
UN2811
Off-white
1g
89976-12-5
98%
MFCD01631684
2506013
1-Methyl-4-nitro-2-(trifluoromethyl)benzene
Moisture Sensitive
Videos
SDS