missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-hydroxytryptamine hydrochloride, Tocris Bioscience™

5-HT3 agonist/potent 5-HT6 ligand

Brand:  Tocris (Bio-Techne) 055850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437763

  • £349.81 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

5-HT3 agonist (Ki = 1200 nM) and potent 5-HT6 ligand (Ki = 46 nM).
Specifications

Chemical Identifiers

845861-49-6
226.704
2-methylserotonin.hcl, 2-methyl-5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-2-methyl-1h-indol-5-ol hydrochloride, 2-methyl-5-ht hydrochloride
3-(2-aminoethyl)-2-methyl-1H-indol-5-ol;hydrochloride
C11H15ClN2O
ZNVRHQPXSBWLQP-UHFFFAOYSA-N
11492320
CC1=C(C2=C(N1)C=CC(=C2)O)CCN.Cl

Specifications

2-Methyl-5-hydroxytryptamine hydrochloride
845861-49-6
2-methylserotonin.hcl, 2-methyl-5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-2-methyl-1h-indol-5-ol hydrochloride, 2-methyl-5-ht hydrochloride
CC1=C(C2=C(N1)C=CC(=C2)O)CCN.Cl
226.704
226.71
50mg
C11H15ClN2O
ZNVRHQPXSBWLQP-UHFFFAOYSA-N
3-(2-aminoethyl)-2-methyl-1H-indol-5-ol;hydrochloride
11492320
>98%
Videos