missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid, 98%

Brand:  Alfa Aesar™ H64592.14

Code : NEW

Additional Details : CAS Number : 117724-63-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15439006

  • £298.92 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
211.16
9848326
117724-63-7
MFCD00173295

Specifications

2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
25g
Solid
211.16
9848326
186° to 187°C
117724-63-7
98%
MFCD00173295
Videos
SDS