missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid, 98%

Brand:  Alfa Aesar™ H64592.06

Code : NEW

Additional Details : CAS Number : 117724-63-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429006

  • £59.78 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
211.16
9848326
117724-63-7
MFCD00173295

Specifications

2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
5g
Solid
211.16
9848326
186° to 187°C
117724-63-7
98%
MFCD00173295
Videos
SDS