missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog Number p-7053972
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
67169-22-6
C8H8F3N
175.154
MFCD03701068
PAXQXJDYVORMOO-UHFFFAOYSA-N
2-methyl-4-trifluoromethyl aniline, 4-amino-3-methylbenzotrifluoride, benzenamine, 2-methyl-4-trifluoromethyl, 2-amino-5-trifluoromethyl toluene, pubchem4492, 2-amino-5-trifluoromethyltoluene, 2-methyl-4 trifluoromethyl benzenamine, 2-methyl4-trifluoromethyl benzenamine, 2-methyl-4-trifluoromethyl benzenamine
13750437
2-methyl-4-(trifluoromethyl)aniline
CC1=C(C=CC(=C1)C(F)(F)F)N
This item is not returnable. View return policy
67169-22-6
C8H8F3N
175.154
MFCD03701068
PAXQXJDYVORMOO-UHFFFAOYSA-N
2-methyl-4-trifluoromethyl aniline, 4-amino-3-methylbenzotrifluoride, benzenamine, 2-methyl-4-trifluoromethyl, 2-amino-5-trifluoromethyl toluene, pubchem4492, 2-amino-5-trifluoromethyltoluene, 2-methyl-4 trifluoromethyl benzenamine, 2-methyl4-trifluoromethyl benzenamine, 2-methyl-4-trifluoromethyl benzenamine
13750437
2-methyl-4-(trifluoromethyl)aniline
CC1=C(C=CC(=C1)C(F)(F)F)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
UN Number UN2811
Formula Weight 175.15
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'