missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3881789
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
117724-63-7
C6H4F3NO2S
211.16
MFCD00173295
REKJPVUFKQYMHW-UHFFFAOYSA-N
2-methyl-4-trifluoromethyl thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethylthiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-trifluoromethyl, 5-thiazolecarboxylic acid, 2-methyl-4-trifluoromethyl, pubchem11380, acmc-209yjt, methyltrifluoromethylthiazolecarboxylicacid, 2-methyl-4-trifluoromethyl thiazole-5-carboxylicacid
1486080
2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
CC1=NC(=C(S1)C(O)=O)C(F)(F)F
This item is not returnable. View return policy
117724-63-7
C6H4F3NO2S
211.16
MFCD00173295
REKJPVUFKQYMHW-UHFFFAOYSA-N
2-methyl-4-trifluoromethyl thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethylthiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-trifluoromethyl, 5-thiazolecarboxylic acid, 2-methyl-4-trifluoromethyl, pubchem11380, acmc-209yjt, methyltrifluoromethylthiazolecarboxylicacid, 2-methyl-4-trifluoromethyl thiazole-5-carboxylicacid
1486080
2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
CC1=NC(=C(S1)C(O)=O)C(F)(F)F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 211.16
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'