missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-4-trifluoromethoxy-DL-phenylalanine, 97%

Brand:  Alfa Aesar™ H34333.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447267

  • £120.10 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-4-trifluoromethoxy-DL-phenylalanine
263.21
MFCD22201062

Specifications

2-Methyl-4-trifluoromethoxy-DL-phenylalanine
97%
MFCD22201062
Solid
263.21
1g
Videos
SDS