missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-piperazinoquinoline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
IUPAC Name | 2-methyl-4-piperazin-1-ylquinoline |
Formula Weight | 227.31 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methyl-4-piperazinoquinoline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'