missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenyl-1,3-thiazole-5-carboxylic acid, 97%, Maybridge

Brand:  Maybridge CC42201EA

Code : 31

Additional Details : CAS Number : 32002-72-5 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10757695

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2-Methyl-4-phenyl-1, 3-thiazole-5-carboxylic acid
97%
219.26
97%
C11H9NO2S

Specifications

2-Methyl-4-phenyl-1, 3-thiazole-5-carboxylic acid
32002-72-5
97%
C11H9NO2S
10g
Amber Glass Bottle
97%
219.26
Videos
SDS