missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole, 97%, Maybridge™

£58.00 - £143.00

Chemical Identifiers

CAS 857283-68-2
Molecular Formula C16H20BNO2S
Molecular Weight (g/mol) 301.211
MDL Number MFCD08060513
InChI Key XCAUOIGPROQXRG-UHFFFAOYSA-N
Synonym 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazole, 2-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1,3-thiazole, thiazole,2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
PubChem CID 18525749
IUPAC Name 2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CSC(=N3)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10622485
SDS View Documents
Maybridge
CC40339CB
250mg Amber Glass Bottle
£58.27
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Please to purchase this item. Need a web account? Register with us today!
10114243
SDS View Documents
Maybridge
CC40339DA
1g Amber Glass Bottle
£143.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Please to purchase this item. Need a web account? Register with us today!
11380497
SDS View Documents
Maybridge
CC40339CB
250mg Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

857283-68-2
301.211
XCAUOIGPROQXRG-UHFFFAOYSA-N
18525749
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CSC(=N3)C
C16H20BNO2S
MFCD08060513
2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazole, 2-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1,3-thiazole, thiazole,2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole

Specifications

2-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
250mg
96.5
97%
MFCD08060513
XCAUOIGPROQXRG-UHFFFAOYSA-N
2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
18525749
97%
Amber Glass Bottle
857283-68-2
100.0
C16H20BNO2S
2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazole, 2-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1,3-thiazole, thiazole,2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CSC(=N3)C
301.211
301.22
SDS
Product Certifications