missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-4-(2-pyridyl)thiazole, 97%

Brand:  Alfa Aesar™ H58878.09

Code : NEW

Additional Details : CAS Number : 14384-71-5 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15415068

  • £731.60 / 10g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-4-(2-pyridyl)thiazole
176.24
1565687
14384-71-5
MFCD22666465

Specifications

2-Methyl-4-(2-pyridyl)thiazole
14384-71-5
97%
MFCD22666465
180° to 185°C
10g
Powder
176.24
1565687
Pale brown
Videos
SDS