missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(trifluoromethyl)aniline, 97%, ACROS Organics™

Brand:  Acros Organics 263800050

Code : 29

Additional Details : CAS Number : 54396-44-0 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11985211

  • £196.20 / 5g


Estimated Shipment
12-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Methyl-3-(trifluoromethyl)aniline
96.0
97%
C8H8F3N
3-(Trifluoromethyl)-o-toluidine
54396-44-0
100.0
97%
175.16

Specifications

2-Methyl-3-(trifluoromethyl)aniline
108.0° to 110.0°C
39.0° to 41.0°C
54396-44-0
100.0
1.4825
97%
C8H8F3N
3-(Trifluoromethyl)-o-toluidine
96°C
White
5g
96.0
Authentic
Crystalline Powder
97%
175.16
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methyl-3-(trifluoromethyl)aniline

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS