missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-3-nitroaniline, 98%

Brand:  Alfa Aesar™ A10347.22

Code : NEW

Additional Details : CAS Number : 603-83-8 Weight : 0.10000kg Transport : UN number : 2660 Chem class : 1 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11414143

  • £48.10 / 100g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-3-nitroaniline
C7H8N2O2
MFCD00007731
388393
603-83-8
152.15
UN2660
2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine

Specifications

2-Methyl-3-nitroaniline
98%
152.15
UN2660
2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine
305°C
Characteristic
603-83-8
C7H8N2O2
MFCD00007731
388393
88° to 91°C
100g
Videos
SDS