missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-3-furyl)methylamine 97+%, Maybridge

Brand:  Maybridge CC55413DA

Code : 31

Additional Details : CAS Number : 35801-15-1 Weight : 0.05100kg Transport : UN number : 2733 Chem class : 3 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 10123824

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(2-Methyl-3-furyl)methylamine
97%
111.14
97%
C6H9NO

Specifications

(2-Methyl-3-furyl)methylamine
35801-15-1
97%
C6H9NO
1g
Amber Glass Bottle
97%
111.14
Videos
SDS