missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Methyl-3-furyl)methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Synonym | 2-methylfuran-3-yl methanol, 2-methyl-3-furyl methanol, 2-methyl-3-furanmethanol, 3-hydroxymethyl-2-methylfuran, 3-furanmethanol, 2-methyl, 2-methyl-furan-3-yl-methanol, 3-furanmethanol,2-methyl, 2-methylfur-3-yl methanol, 2-methyl-3-furyl methan-1-ol |
Formula Weight | 112.13 |
Percent Purity | 97% |
Chemical Name or Material | (2-Methyl-3-furyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'