missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-furanethiol, ca.90%, balance oxidized compound, ACROS Organics™

Brand:  Acros Organics 275390050

Code : 29

Additional Details : CAS Number : 28588-74-1 Weight : 0.05500kg Transport : UN number : 3071 Chem class : 1 Pack group : II


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10539443

  • £148.50 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Methyl-3-furanethiol
88.0
≥88%
C5H6OS
MFCD00010280
28588-74-1
100.0
ca.90%
114.17

Specifications

2-Methyl-3-furanethiol
5g
88.0
1.1450g/mL
Glass bottle
important note: this substance readily oxidizes in air to form bis(2-methyl-3-fyryl)disulphide
≥88%
C5H6OS
MFCD00010280
Clear to hazy light pink to light orange
28588-74-1
100.0
Authentic
1.5100 to 1.5500 (20°C, 589nm)
Liquid
ca.90%
114.17
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methyl-3-furanethiol, balance oxidized compound

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS