missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-buten-2-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-160665
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
115-18-4
C5H10O
86.13
MFCD00004470
HNVRRHSXBLFLIG-UHFFFAOYSA-N
2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
8257
2-methylbut-3-en-2-ol
CC(C)(C=C)O
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
115-18-4
C5H10O
86.13
MFCD00004470
HNVRRHSXBLFLIG-UHFFFAOYSA-N
2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
8257
2-methylbut-3-en-2-ol
CC(C)(C=C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 98.0°C to 99.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC(CH3)2OH
Refractive Index 1.4160 to 1.4180
Quantity 250 mL
Beilstein 01, 444
Specific Gravity 0.82
Solubility Information Solubility in water: 100g/L (20 c)
Formula Weight 86.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-buten-2-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-3-buten-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'