missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2H-indazole-3-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-4510663
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
34252-54-5
C9H8N2O
160.18
MFCD00839530
SMWLSZQHLMZHHV-UHFFFAOYSA-N
2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl
818117
2-methylindazole-3-carbaldehyde
CN1N=C2C=CC=CC2=C1C=O
This item is not returnable. View return policy
34252-54-5
C9H8N2O
160.18
MFCD00839530
SMWLSZQHLMZHHV-UHFFFAOYSA-N
2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl
818117
2-methylindazole-3-carbaldehyde
CN1N=C2C=CC=CC2=C1C=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 2-Methyl-2H-indazole-3-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'