missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propanethiol, 99%, Thermo Scientific Chemicals

Catalog Number 10467730
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
75-66-1
C4H10S
90.18
MFCD00004857
WMXCDAVJEZZYLT-UHFFFAOYSA-N
2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
6387
2-methylpropane-2-thiol
CC(C)(C)S
This item is not returnable. View return policy
75-66-1
C4H10S
90.18
MFCD00004857
WMXCDAVJEZZYLT-UHFFFAOYSA-N
2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
6387
2-methylpropane-2-thiol
CC(C)(C)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.1°C
Color Colorless
Density 0.8000g/mL
Boiling Point 62°C to 65°C
Flash Point -24°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSH
Refractive Index 1.4220 to 1.424
Quantity 500 mL
Beilstein 01, 383
Merck Index 15, 1582
Specific Gravity 0.8
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in liquid h2s,very soluble in alcohol and ether
Viscosity 0.638 mPa.s (20°C)
Formula Weight 90.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-propanethiol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-propanethiol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'