missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentenal, 97%, Thermo Scientific Chemicals

Catalog Number p-3758455
18.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-36-9
C6H10O
98.15
MFCD00006978
IDEYZABHVQLHAF-GQCTYLIASA-N
2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
5319754
(E)-2-methylpent-2-enal
CCC=C(C)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-36-9
C6H10O
98.15
MFCD00006978
IDEYZABHVQLHAF-GQCTYLIASA-N
2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
5319754
(E)-2-methylpent-2-enal
CCC=C(C)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 134.0°C to 136.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH=C(CH3)CHO
Refractive Index 1.4470 to 1.4550
Quantity 25 g
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble
Formula Weight 98.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-pentenal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-pentenal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'